LIF372155493 Life Chemicals Inc.

5-(4-ethoxyphenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD02084772
  • Cas Number : 84159-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart