LIF372155494 Life Chemicals Inc.

5-(3,4-dimethoxyphenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD01465889
  • Cas Number : 23269-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart