LIF372155511 Life Chemicals Inc.

4-(4-methoxy-3-methylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD00987014
  • Cas Number : 92388-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart