LIF372155512 Life Chemicals Inc.

[3-(1,3-benzothiazol-2-yl)-6-methyl-4,5,6,7-tetrahydro-1-benzothien-2-yl]amine

  • MDL number MFCD11986727
  • Cas Number : 1105194-23-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart