LIF372155513 Life Chemicals Inc.

5,6-Dihydro-4H-cyclopentathiazol-2-ylamine

  • MDL number MFCD00159774
  • Cas Number : 53051-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart