LIF372155516 Life Chemicals Inc.

Methyl 3-Amino-5-(4-phenyl)thiophene-2-carboxylate

  • MDL number MFCD00068161
  • Cas Number : 100063-22-7
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart