LIF372155524 Life Chemicals Inc.

acenaphtho[1,2-d][1,3]thiazol-8-amine

  • MDL number MFCD06660593
  • Cas Number : 7496-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart