LIF372155533 Life Chemicals Inc.

ethyl 2-amino-3-(1,3-benzothiazol-2-yl)-4,7-dihydrothieno[2,3-c]pyridine-6(5H)-carboxylate

  • MDL number MFCD11986730
  • Cas Number : 1105194-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart