LIF372155539 Life Chemicals Inc.

5-(3-methoxyphenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09028377
  • Cas Number : 1673-44-5
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart