LIF372155543 Life Chemicals Inc.

6-methyl-2-piperazin-1-yl-1,3-benzothiazole

  • MDL number MFCD10008524
  • Cas Number : 1018498-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart