LIF372155548 Life Chemicals Inc.

6-Fluoro-2-piperazin-1-yl-1,3-benzothiazole

  • MDL number MFCD09743163
  • Cas Number : 401567-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart