LIF372155559 Life Chemicals Inc.

4,7-dimethoxy-2-piperazin-1-yl-1,3-benzothiazole

  • MDL number MFCD11986735
  • Cas Number : 1105194-52-2
  • Molecular Weight : 279.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart