LIF372155566 Life Chemicals Inc.

4-(methylthio)-1,3-benzothiazol-2-amine

  • MDL number MFCD11986737
  • Cas Number : 1105194-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart