LIF372155567 Life Chemicals Inc.

5-(3,4-dimethylphenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09807228
  • Cas Number : 1016528-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart