LIF372155568 Life Chemicals Inc.

7-Chloro-4-methoxy-benzothiazol-2-ylamine

  • MDL number MFCD09743170
  • Cas Number : 67618-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart