LIF372155570 Life Chemicals Inc.

4-(methylthio)-2-piperazin-1-yl-1,3-benzothiazole

  • MDL number MFCD11986738
  • Cas Number : 1105194-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart