LIF372155576 Life Chemicals Inc.

5-[2-(phenylthio)ethyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD11986743
  • Cas Number : 1105194-76-0
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart