LIF372155581 Life Chemicals Inc.

6-ethyl-N-(pyridin-2-ylmethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD11986747
  • Cas Number : 1105194-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart