LIF372155586 Life Chemicals Inc.

6-bromo-N-(pyridin-2-ylmethyl)benzo[d]thiazol-2-amine

  • MDL number MFCD09743200
  • Cas Number : 941867-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart