LIF372155593 Life Chemicals Inc.

5,7-dimethyl-N-(pyridin-2-ylmethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD11986754
  • Cas Number : 1105191-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart