LIF372155595 Life Chemicals Inc.

4,7-dimethyl-N-(pyridin-2-ylmethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD11986756
  • Cas Number : 1105191-30-7
  • Molecular Weight : 269.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart