LIF372155601 Life Chemicals Inc.

N-benzyl-4-methyl-1,3-benzothiazol-2-amine

  • MDL number MFCD03653032
  • Cas Number : 345621-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart