LIF372155607 Life Chemicals Inc.

N-benzyl-4,7-dimethyl-1,3-benzothiazol-2-amine

  • MDL number MFCD16631688
  • Cas Number : 1177308-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart