LIF372155618 Life Chemicals Inc.

4-methoxy-N-(pyridin-3-ylmethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD09743207
  • Cas Number : 941867-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart