LIF372155632 Life Chemicals Inc.

4,7-dimethoxy-N-(pyridin-3-ylmethyl)benzo[d]thiazol-2-amine

  • MDL number MFCD11986772
  • Cas Number : 1105192-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart