LIF372155640 Life Chemicals Inc.

5-chlorothiophene-2-carbonyl chloride

  • MDL number MFCD00043886
  • Cas Number : 42518-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart