LIF372155662 Life Chemicals Inc.

7-chloro-4-methyl-N-(pyridin-4-ylmethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD16631691
  • Cas Number : 1177306-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart