LIF372155669 Life Chemicals Inc.

4,5-dimethyl-N-(2-morpholin-4-ylethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD16631697
  • Cas Number : 1177298-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart