LIF372155672 Life Chemicals Inc.

4,6-difluoro-N-[3-(1H-imidazol-1-yl)propyl]-1,3-benzothiazol-2-amine

  • MDL number MFCD16631700
  • Cas Number : 1177275-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart