LIF372155682 Life Chemicals Inc.

6-ethyl-N-[3-(1H-imidazol-1-yl)propyl]-1,3-benzothiazol-2-amine

  • MDL number MFCD16631710
  • Cas Number : 1177285-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart