LIF372155683 Life Chemicals Inc.

N-[3-(1H-imidazol-1-yl)propyl]-6-methoxy-1,3-benzothiazol-2-amine

  • MDL number MFCD09031548
  • Cas Number : 862820-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart