LIF372155693 Life Chemicals Inc.

N'-(5-methoxy-1,3-benzothiazol-2-yl)-N,N-dimethylethane-1,2-diamine

  • MDL number MFCD16631719
  • Cas Number : 1177284-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart