LIF372155699 Life Chemicals Inc.

5-(5-bromo-2-furyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09816167
  • Cas Number : 1016711-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart