LIF372155710 Life Chemicals Inc.

5-[4-(isopropylsulfonyl)phenyl]-1,3,4-oxadiazol-2-amine

  • MDL number MFCD16631726
  • Cas Number : 1177318-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart