LIF372155722 Life Chemicals Inc.

2-methyl-2,4,5,6-tetrahydrocyclopenta[c]pyrazol-3-amine

  • MDL number MFCD08444199
  • Cas Number : 885529-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart