LIF372155749 Life Chemicals Inc.

5-(chloromethyl)-1-(4-chlorophenyl)-1H-tetrazole

  • MDL number MFCD08061133
  • Cas Number : 858483-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart