LIF372155764 Life Chemicals Inc.

1,2,3,4-Tetrahydropyrrolo[1,2-a]pyrazine-1-carboxylic acid

  • MDL number MFCD16653012
  • Cas Number : 1214086-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart