LIF372155770 Life Chemicals Inc.

Ethyl 3-methyl-1,2,3,4-tetrahydro-3-quinolinecarboxylate

  • MDL number MFCD16653016
  • Cas Number : 1105193-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart