LIF372155773 Life Chemicals Inc.

3-(3-methyl-1,2-oxazol-5-yl)propan-1-ol

  • MDL number MFCD04972527
  • Cas Number : 105658-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart