LIF372155785 Life Chemicals Inc.

3-hydroxy-4-nitrobenzonitrile

  • MDL number MFCD06657032
  • Cas Number : 18495-15-3
  • Molecular Weight : 164.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart