LIF372155841 Life Chemicals Inc.

methyl 1-isopropyl-5-oxopyrrolidine-3-carboxylate

  • MDL number MFCD11053005
  • Cas Number : 59857-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart