LIF372155854 Life Chemicals Inc.

[1-(3-Fluorophenyl)-3-pyrrolidinyl]methanamine

  • MDL number MFCD09939491
  • Cas Number : 1017428-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart