LIF372155953 Life Chemicals Inc.

5-Amino-1-(4-bromophenyl)-1H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD09796206
  • Cas Number : 929837-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart