LIF372155962 Life Chemicals Inc.

5-Amino-1-(3-chloro-4-methylphenyl)-1H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD07401841
  • Cas Number : 887689-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart