LIF372155971 Life Chemicals Inc.

1-(2-methoxyphenyl)-1H-imidazole-2-thiol

  • MDL number MFCD02948337
  • Cas Number : 51581-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart