LIF372155988 Life Chemicals Inc.

1-(1,3-benzodioxol-5-ylmethyl)-1H-imidazole-2-thiol

  • MDL number MFCD11986805
  • Cas Number : 1105195-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart