LIF372156078 Life Chemicals Inc.

2-chloro-3-(propan-2-yloxy)pyridine

  • MDL number MFCD16653100
  • Cas Number : 1105190-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart