LIF372156080 Life Chemicals Inc.

2-chloro-3-[(2-fluorobenzyl)oxy]pyridine

  • MDL number MFCD11986848
  • Cas Number : 1105190-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart