LIF372156081 Life Chemicals Inc.

2-Chloro-3-[(4-fluorobenzyl)oxy]pyridine

  • MDL number MFCD11986849
  • Cas Number : 1105190-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart