LIF372156103 Life Chemicals Inc.

5-amino-1-(2-fluorobenzyl)-1H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD11986861
  • Cas Number : 93416-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart